Temozolomide ≥98%
Lieferant: ENZO LIFE SCIENCES
Synonyme: 3,4-Dihydro-3-methyl-4-oxoimidazo[5,1-d][1,2,3,5]tetrazine-8-carboxamide, 3-Methyl-4-oxo-3,4-dihydroimidazo[5,1-d][1,2,3,5]tetrazine-8-carboxamide
ALX420044M025 ALX420044M100 ALX420044M500
ENZOALX420044M025EA 147 EUR
ENZOALX420044M025 ENZOALX420044M100 ENZOALX420044M500
Temozolomide ≥98%
Temozolomide
AGT inhibitor
Antitumor and antiangiogenic compound. DNA alkylating agent. Induces G2/M arrest and apoptosis through adduction of a methyl group to O6 position of guanine in genomic DNA. Inactivates DNA repair protein O(6)-alkylguanine DNA alkyltransferase (AGT) in base excision repair (BER) pathway. Chemotherapeutic agent. Modifies drug resistance.
Formel: C₆H₆N₆O₂
Molecular Weight: 194.15 g/mol
MDL: MFCD00866492
CAS-Nummer: 85622-93-1