Purmorphamine ≥98% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Achtung
Synonyme: 2-(1-Naphthoxy)-6-(4-morpholinoanilino)-9-cyclohexylpurine, 9-Cyclohexyl-N-(4-morpholinophenyl)-2-(naphthalen-1-yloxy)-9H-purin-6-amine
ALX420045M001 ALX420045M005 ALX420045M025
ENZOALX420045M001EA 193 EUR
ENZOALX420045M001 ENZOALX420045M005 ENZOALX420045M025
Purmorphamine ≥98% (durch HPLC)
Purmorphamine
Smoothened activator.
Cell permeable purine compound. Induces osteoblast differentiation. Agonist of smoothened by activation of the Hedgehog (Hh) pathway. Also shown to induce transdifferentiation in pre-adipocytes and myoblasts. Binds to and activates 7-transmembrane receptor of Hh.
Soluble in DMSO (10 mM).
Formel: C₃₁H₃₂N₆O₂
Molecular Weight: 520.637 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD09037557
CAS-Nummer: 483367-10-8
Bestellen
Spezifikation Testergebnisse
Purity ≥98% (HPLC)