MAHMA NONOate ≥98%

Lieferant: ENZO LIFE SCIENCES
Achtung

ALX4300155005 ALX430015M005 ALX430015M025
ENZOALX4300155005EA 227 EUR
ENZOALX4300155005 ENZOALX430015M005 ENZOALX430015M025
MAHMA NONOate ≥98%
MAHMA NONOate

Nitric oxide (NO) donor.


The half-life time of MAHMA NONOate is 2.7 min. at 22 °C in 0.1 M phosphate buffer, pH 7.4. Decomposition of NONOates is nearly instantaneous at pH 5.0.


Soluble in water. Relatively stable in alkaline solution.


Bitte beachten Protect from light and moisture.

Formel: C₈H₂₀N₄O₂
Molecular Weight: 204,3 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 146724-86-9

Bestellen

Spezifikation Testergebnisse

Purity ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR