PAPA NONOate ≥97%

Lieferant: ENZO LIFE SCIENCES
Achtung

ALX4300165005 ALX430016M005 ALX430016M025
ENZOALX4300165005EA 227 EUR
ENZOALX4300165005 ENZOALX430016M005 ENZOALX430016M025
PAPA NONOate ≥97%
PAPA NONOate

Nitric oxide (NO) donor.


The half-life time of PAPA NONOate is 76.6 min at 22 °C in 0.1 M phosphate buffer, pH 7.4. Decomposition of NONOates is nearly instantaneous at pH 5.0.


Soluble in water or methanol. Relatively stable in alkaline solution.


Bitte beachten Protect from moisture. Keep under inert gas.

Formel: C₆H₁₆N₄O₂
Molecular Weight: 176,2 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 146672-58-4

Bestellen

Spezifikation Testergebnisse

Purity ≥97%
Identity Determined by IR

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR