BPMO ≥99% (durch 1H-NMR), hochrein
Lieferant: ENZO LIFE SCIENCES
Synonyme: 2-(tert-Butoxycarbonyl)-2-methyl-3,4-dihydro-2H-pyrrole 1-oxide, 5-tert-Butoxycarbonyl-5-methyl-1-pyrroline-N-oxide, BMPO
ALX430141M010 ALX430141M050
ENZOALX430141M010EA 200 EUR
ENZOALX430141M010 ENZOALX430141M050
BPMO ≥99% (durch 1H-NMR), hochrein
BMPO
Nitrone spin trap for the specific in vivo or in vitro detection of short-lived superoxide, hydroxyl and thiyl radicals.
Forms distinguishable adducts which can be measured by EPR spectroscopy. Unlike with DMPO, the superoxide adduct does not decay into a hydroxyl adduct and it has a much longer half-life (t1/2=23 min). Low paramagnetic impurities. No further purification required.
Soluble in water.
Formel: C₁₀H₁₇NO₃
Molecular Weight: 199.3 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD32196548
CAS-Nummer: 387334-31-8
Bestellen
Spezifikation Testergebnisse
Purity (1H-NMR, EPR) ≥99%