Lamotrigin ≥98%
Lieferant: ENZO LIFE SCIENCES
Synonyme:
6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine, 3,5-Diamino-6-(2,3-dichlorophenyl)-1,2,4-triazine
ALX430160M025
ALX430160M100
ALX430160M500
ENZOALX430160M025EA
245
EUR
ENZOALX430160M025
ENZOALX430160M100
ENZOALX430160M500
Lamotrigin ≥98%
Lamotrigine
Na⁺ channel blocker
Lamotrigine blocks voltage-gated sodium channels. Inhibits the release of glutamate. Neuroprotective. Anticonvulsant.
Formel:
C₉H₇Cl₂N₅ Molecular Weight: 256.09 g/mol |
MDL:
MFCD00865333 CAS-Nummer: 84057-84-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...