LY-83583 ≥95%

Lieferant: ENZO LIFE SCIENCES

Synonyme: 6-Anilinoquinoline-5,8-quinone

ALX550002M005 ALX550002M025
ENZOALX550002M005EA 124 EUR
ENZOALX550002M005 ENZOALX550002M025
LY-83583 ≥95%
LY-83583

Guanylyl cyclase inhibitor


Competitive inhibitor of soluble guanylyl cyclase (sGC) (IC50=2µM) and of cGMP production. Neurotoxin. Blocks antigen-induced leukotriene release and reduces platelet-endothelial cell adhesion. Blocks acethylcholine-induced vasorelaxation.


Synthetic.

Formel: C₁₅H₁₀N₂O₂
Molecular Weight: 250,25 g/mol
CAS-Nummer: 91300-60-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR