Luzindole ≥96%
Lieferant: ENZO LIFE SCIENCES
Synonyme:
N-Acetyl-2-benzyl-tryptamine
ALX550184M005
ALX550184M025
ENZOALX550184M005EA
101
EUR
ENZOALX550184M005
ENZOALX550184M025
Luzindole ≥96%
Luzindole
Competitive melatonin receptor antagonist.
Formel:
C₁₉H₂₀N₂O Molecular Weight: 292,382 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00672498 CAS-Nummer: 117946-91-5 |
Spezifikation Testergebnisse
Purity | ≥96% |
Appearance | White to off-white crystalline solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...