Serotonin Hydrochlorid
Lieferant: ENZO LIFE SCIENCES
Synonyme: 5-Hydroxytryptamine Hydrochloride
ALX550328G001 ALX550328M050
ENZOALX550328G001EA 317 EUR
ENZOALX550328G001 ENZOALX550328M050
Serotonin Hydrochlorid
Serotonin Hydrochlorid
Formel: C₁₀H₁₃ClN₂O
Molecular Weight: 212.68 g/mol
MDL: MFCD00012686
CAS-Nummer: 153-98-0
UN: 2811
ADR: 6.1 III