Flumazenil ≥98%
Lieferant: ENZO LIFE SCIENCES
Synonyme:
Ethyl 8-fluoro-5-methyl-6-oxo-5,6-dihydro-4H-benzo[f]imidazo[1,5-a][1,4]diazepine-3-carboxylate
BMLB1000100
ENZOBMLB1000100EA
371
EUR
ENZOBMLB1000100
Flumazenil ≥98%
Flumazenil
Benzodiazepine receptor ligand.
Benzodiazepine antagonist. Reverses all the effects of benzodiazepine agonists and inverse agonists.
Soluble in ethanol (5 mg/ml warm) or DMSO (25 mg/ml warm).
Formel:
C₁₅H₁₄FN₃O₃ Molecular Weight: 303,29 g/mol |
MDL:
MFCD00242764 CAS-Nummer: 78755-81-4 |
Spezifikation Testergebnisse
Purity | ≥98% (Assay) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...