Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (APC (Allophycocyanin)/iFluor® 700)

Lieferant: AAT BIOQUEST
101921F0 101921F1 101921F2
AATB101921F0EA 319 EUR
AATB101921F0 AATB101921F1 AATB101921F2
Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (APC (Allophycocyanin)/iFluor® 700)
Antikörper
HIB19 is an anti-human monoclonal antibody that recognises the CD19 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: HIB19
Conjugation: APC (Allophycocyanin)/iFluor® 700
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR