Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (APC (Allophycocyanin)/iFluor® 700)

Lieferant: AAT BIOQUEST
103401F0 103401F1 103401F2
AATB103401F0EA 319 EUR
AATB103401F0 AATB103401F1 AATB103401F2
Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (APC (Allophycocyanin)/iFluor® 700)
Antikörper
4H11 is an anti-human monoclonal antibody that recognises the CD34 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 4H11
Conjugation: APC (Allophycocyanin)/iFluor® 700
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR