Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (APC (Allophycocyanin)/iFluor® 750)
Lieferant: AAT BIOQUEST
104411G0 104411G1 104411G2
AATB104411G0EA 319 EUR
AATB104411G0 AATB104411G1 AATB104411G2
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (APC (Allophycocyanin)/iFluor® 750)
Antikörper
IM7 is an anti-human/mouse monoclonal antibody that targets the CD44 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Bitte beachten For research use only.
Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: IM7
Conjugation: APC (Allophycocyanin)/iFluor® 750
Epitope:
Host: Rat
Isotype:
Reactivity: Human