Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (iFluor® 610)
Lieferant: AAT BIOQUEST
100810D0 100810D1
AATB100810D0EA 371 EUR
AATB100810D0 AATB100810D1
Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (iFluor® 610)
Antikörper
SK1 is an anti-human monoclonal antibody that is specific for the CD8a antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Bitte beachten For research use only.
Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: SK1
Conjugation: iFluor® 610
Epitope:
Host: Mouse
Isotype:
Reactivity: Human