Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (iFluor® 660)

Lieferant: AAT BIOQUEST
100810G0 100810G1
AATB100810G0EA 371 EUR
AATB100810G0 AATB100810G1
Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (iFluor® 660)
Antikörper
SK1 is an anti-human monoclonal antibody that is specific for the CD8a antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: SK1
Conjugation: iFluor® 660
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR