Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (mFluor™ Red 780)

Lieferant: AAT BIOQUEST
101610W0 101610W1
AATB101610W0EA 371 EUR
AATB101610W0 AATB101610W1
Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (mFluor™ Red 780)
Antikörper
The 3G8 monoclonal antibody recognises human CD16a, a 50 to 65 kDa transmembrane protein typically found on the surface of macrophages, natural killer cells, dendritic cells, monocytes and neutrophils.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD16a
Clonality: Monoclonal
Clone: 3G8
Conjugation: mFluor™ Red 780
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR