Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (PE (Phycoerythrin)/iFluor® 594)

Lieferant: AAT BIOQUEST
101921Y0 101921Y1 101921Y2
AATB101921Y0EA 319 EUR
AATB101921Y0 AATB101921Y1 AATB101921Y2
Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (PE (Phycoerythrin)/iFluor® 594)
Antikörper
HIB19 is an anti-human monoclonal antibody that recognises the CD19 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: HIB19
Conjugation: PE (Phycoerythrin)/iFluor® 594
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR