Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (PE (Phycoerythrin)/iFluor® 647)
Lieferant: AAT BIOQUEST
101921Q0 101921Q1 101921Q2
AATB101921Q0EA 319 EUR
AATB101921Q0 AATB101921Q1 AATB101921Q2
Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (PE (Phycoerythrin)/iFluor® 647)
Antikörper
HIB19 is an anti-human monoclonal antibody that recognises the CD19 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Bitte beachten For research use only.
Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: HIB19
Conjugation: PE (Phycoerythrin)/iFluor® 647
Epitope:
Host: Mouse
Isotype:
Reactivity: Human