Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (iFluor® 610)

Lieferant: AAT BIOQUEST
100520D0 100520D1
AATB100520D0EA 371 EUR
AATB100520D0 AATB100520D1
Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (iFluor® 610)
Antikörper
UCHT2 is an anti-human monoclonal antibody that is specific for the CD5 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: iFluor® 610
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR