Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (iFluor® 800)

Lieferant: AAT BIOQUEST
116100N0 116100N1
AATB116100N0EA 371 EUR
AATB116100N0 AATB116100N1
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (iFluor® 800)
Antikörper
HP-3G10 is an anti-human monoclonal antibody that recognises the CD161 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: iFluor® 800
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR