Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (iFluor® 710)

Lieferant: AAT BIOQUEST
104410K0 104410K1
AATB104410K0EA 371 EUR
AATB104410K0 AATB104410K1
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (iFluor® 710)
Antikörper
IM7 is an anti-human/mouse monoclonal antibody that targets the CD44 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD44 antigen
Clonality: Monoclonal
Clone: IM7
Conjugation: iFluor® 710
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR