Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (mFluor™ Violet 540)

Lieferant: AAT BIOQUEST
10141120 10141121
AATB10141120EA 371 EUR
AATB10141120 AATB10141121
Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (mFluor™ Violet 540)
Antikörper
61D3 is an anti-human monoclonal antibody that targets the CD14 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD14 antigen
Clonality: Monoclonal
Clone: 61D3
Conjugation: mFluor™ Violet 540
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR