Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (PE/iFluor® 594)

Lieferant: AAT BIOQUEST
101411Y0 101411Y1 101411Y2
AATB101411Y0EA 319 EUR
AATB101411Y0 AATB101411Y1 AATB101411Y2
Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (PE/iFluor® 594)
Antikörper
61D3 is an anti-human monoclonal antibody that targets the CD14 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD14 antigen
Clonality: Monoclonal
Clone: 61D3
Conjugation: PE/iFluor® 594
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR