Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (PE/iFluor® 594)

Lieferant: AAT BIOQUEST
103801Y0 103801Y1 103801Y2
AATB103801Y0EA 319 EUR
AATB103801Y0 AATB103801Y1 AATB103801Y2
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (PE/iFluor® 594)
Antikörper
HIT2 is an anti-human monoclonal antibody that targets the CD38 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD38 antigen
Clonality: Monoclonal
Clone: HIT2
Conjugation: PE/iFluor® 594
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR