Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (PE/iFluor® 594)

Lieferant: AAT BIOQUEST
100511Y0 100511Y1 100511Y2
AATB100511Y0EA 319 EUR
AATB100511Y0 AATB100511Y1 AATB100511Y2
Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (PE/iFluor® 594)
Antikörper
L17F12 is an anti-human monoclonal antibody that is specific for the CD5 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD5 antigen
Clonality: Monoclonal
Clone: L17F12
Conjugation: PE/iFluor® 594
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR