Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (PE/iFluor® 647)

Lieferant: AAT BIOQUEST
101911Q0 101911Q1 101911Q2
AATB101911Q0EA 319 EUR
AATB101911Q0 AATB101911Q1 AATB101911Q2
Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (PE/iFluor® 647)
Antikörper
SJ25C1 is an anti-human monoclonal antibody that forms an immune complex with the CD19 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD19 antigen
Clonality: Monoclonal
Clone: SJ25C1
Conjugation: PE/iFluor® 647
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR