Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (iFluor® 660)

Lieferant: AAT BIOQUEST
100510G0 100510G1
AATB100510G0EA 371 EUR
AATB100510G0 AATB100510G1
Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (iFluor® 660)
Antikörper
L17F12 is an anti-human monoclonal antibody that is specific for the CD5 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: L17F12
Conjugation: iFluor® 660
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR