Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (mFluor™ Green 620)

Lieferant: AAT BIOQUEST
101410U0 101410U1
AATB101410U0EA 371 EUR
AATB101410U0 AATB101410U1
Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (mFluor™ Green 620)
Antikörper
61D3 is an anti-human monoclonal antibody that targets the CD14 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: 61D3
Conjugation: mFluor™ Green 620
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR