Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (mFluor™ Red 780)

Lieferant: AAT BIOQUEST
103800W0 103800W1
AATB103800W0EA 371 EUR
AATB103800W0 AATB103800W1
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (mFluor™ Red 780)
Antikörper
HIT2 is an anti-human monoclonal antibody that targets the CD38 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HIT2
Conjugation: mFluor™ Red 780
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR