Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (PE (Phycoerythrin)/iFluor® 594)

Lieferant: AAT BIOQUEST
102801Y0 102801Y1 102801Y2
AATB102801Y0EA 319 EUR
AATB102801Y0 AATB102801Y1 AATB102801Y2
Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (PE (Phycoerythrin)/iFluor® 594)
Antikörper
CD28.2 is an anti-human monoclonal antibody that forms an immune complex with the CD28 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: CD28.2
Conjugation: PE (Phycoerythrin)/iFluor® 594
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR