Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (PE (Phycoerythrin)/iFluor® 594)

Lieferant: AAT BIOQUEST
100331Y0 100331Y1 100331Y2
AATB100331Y0EA 319 EUR
AATB100331Y0 AATB100331Y1 AATB100331Y2
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (PE (Phycoerythrin)/iFluor® 594)
Antikörper
The SK7 monoclonal antibody reacts with human CD3e, a 20 kD member of the Ig superfamily often found on the surface of t cells, tregs and nkt cells.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: SK7
Conjugation: PE (Phycoerythrin)/iFluor® 594
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR