Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (PE (Phycoerythrin)/iFluor® 647)
Lieferant: AAT BIOQUEST
102801Q0 102801Q1 102801Q2
AATB102801Q0EA 319 EUR
AATB102801Q0 AATB102801Q1 AATB102801Q2
Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (PE (Phycoerythrin)/iFluor® 647)
Antikörper
CD28.2 is an anti-human monoclonal antibody that forms an immune complex with the CD28 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Bitte beachten For research use only.
Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: CD28.2
Conjugation: PE (Phycoerythrin)/iFluor® 647
Epitope:
Host: Mouse
Isotype:
Reactivity: Human