Anti-CD25 Mouse Monoclonal Antibody [clone: BC96] (Biotin)

Lieferant: STEMCELL Technologies
STMC60158BTEA 350 EUR
STMC60158BT
Anti-CD25 Mouse Monoclonal Antibody [clone: BC96] (Biotin)
Antikörper
Mouse monoclonal IgG1 antibody against human, rhesus, cynomolgus CD25, biotin-conjugated.

  • B cells
  • T cells
  • T cells Regulatory

The BC96 antibody reacts with human CD25, a 55 kDa type I integral transmembrane glycoprotein. CD25 is involved in the activation and proliferation of lymphocytes and can be used in the identification of T cell subsets in viral infections like HIV and HTLV. CD25 is present on activated T and B cells, monocytes, macrophages, and regulatory T cells. Expression of CD25, together with CD4 and FOXP3, is considered a phenotypic signature for regulatory T cells. CD25 has a low affinity for its IL-2 ligand but associates with IL-2 receptor β (CD122) and common γ (CD132) chains to form a high affinity IL-2 receptor.

Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD25
Clonality: Monoclonal
Clone: BC96
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Baboon, Chimpanzee, Cynomolgus monkey, Human, Macaque, Rhesus monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR