Mouse IFN-gamma ELISA Kits
Lieferant: STEMCELL Technologies
02020 02021
STMC02020EA 1220 EUR
STMC02020 STMC02021
Mouse IFN-gamma ELISA Kits
Tests ELISAs
For detection and measurement of mouse interferon-gamma.
The Mouse Interferon-gamma (IFN-γ) ELISA Kit is designed for the quantitative detection and measurement of mouse IFN-γ in biological fluids such as serum, plasma, and cell culture supernatants. IFN-γ is primarily secreted by T and NK cells, but is also produced by dendritic cells and macrophages in smaller amounts. IFN-γ is a pleiotropic cytokine that modulates a broad range of immunological functions including antimicrobial and antitumor activity, and increases the surface expression of MHC class I and II proteins.
The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the cytokine. The captured cytokine is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase, which binds the biotinylated antibody. Addition of the chromogenic enzyme substrate 3,3’,5,5’ tetramethylbenzidine (TMB) results in a coloured product with an intensity directly proportional to the concentration of cytokine in the sample. The concentration of the cytokine is determined by comparison to a serial dilution of the cytokine standard analyzed in parallel.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Bestellen
TECHNISCHE DATEN
Environmentally Preferable
Target protein IFN-gamma
Reaktivität der primären Antikörper Mouse
Wirt
Learn more