Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (iFluor® 710)

Lieferant: AAT BIOQUEST
50118 50119
AATB50118EA 153 EUR
AATB50118 AATB50119
Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (iFluor® 710)
Antikörper
Anti-human secondary antibodies have well-characterised specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target.

  • Affinity purified
  • Easily reconstituted

Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence).

Bitte beachten For research use only.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: iFluor® 710
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR