Anti-Maltose Binding Protein Goat Polyclonal Antibody
Lieferant: ANTIBODIES.COM
A121587-15 A121587-600
ANTIA121587-15EA 635 EUR
ANTIA121587-15 ANTIA121587-600
Anti-Maltose Binding Protein Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody to Maltose Binding Protein for WB with samples derived from Escherichia coli and MBP tagged proteins.
  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: Maltose Binding Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Escherichia coli, MBP tagged proteins