Anti-IgG Goat Polyclonal Antibody (iFluor® 800)

Lieferant: AAT BIOQUEST
48002 48003
AATB48002EA 184 EUR
AATB48002 AATB48003
Anti-IgG Goat Polyclonal Antibody (iFluor® 800)
Antikörper
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.

  • Purified and quality-tested for consistent results
  • Suitable for multiple applications
  • Purified through affinity chromatography

iFluor® 800 goat anti-mouse IgG (H+L) conjugate has fluorescence excitation and emission maxima of ∼801 nm and ∼820 nm, respectively.

Bitte beachten Research use only.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: iFluor® 800
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR