Anti-IgG Goat Polyclonal Antibody (mFluor™ UV 375)

Lieferant: AAT BIOQUEST
49560 49561
AATB49560EA 266 EUR
AATB49560 AATB49561
Anti-IgG Goat Polyclonal Antibody (mFluor™ UV 375)
Antikörper
mFluor™ UV375 goat anti-rabbit conjugates are secondary antibodies designed for optimal performance in immunoassay applications, including flow cytometry, immunofluorescence, and confocal microscopy.

  • Purified and quality-tested for consistent results
  • Suitable for multiple applications
  • Purified through affinity chromatography

mFluor™ UV375 is optimally excited by the UV laser and emits maximally at 387 nm.

Bitte beachten Research use only.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: mFluor™ UV 375
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR