Anti-pan Cadherin Mouse Monoclonal Antibody [clone: ABT-MCDH]

Lieferant: ANTIBODIES.COM
A96299-100 A96299-50
ANTIA96299-100EA 475 EUR
ANTIA96299-100 ANTIA96299-50
Anti-pan Cadherin Mouse Monoclonal Antibody [clone: ABT-MCDH]
Antikörper
Mouse monoclonal (ABT-MCDH) antibody to pan Cadherin for IHC and WB with samples derived from Human.

  • Validated Applications: IHC, WB

Type: Primary
Antigen:
Clonality: Monoclonal
Clone: ABT-MCDH
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR