Anti-IgG Goat Polyclonal Antibody (mFluor™ Red 780)

Lieferant: AAT BIOQUEST
49550 49551
AATB49550EA 266 EUR
AATB49550 AATB49551
Anti-IgG Goat Polyclonal Antibody (mFluor™ Red 780)
Antikörper
mFluor™ Red 780 goat anti-mouse conjugates are secondary antibodies designed for optimal performance in immunoassay applications, including flow cytometry, immunofluorescence, and confocal microscopy.

  • Purified and quality-tested for consistent results
  • Suitable for multiple applications
  • Purified through affinity chromatography

mFluor™ Red 780 is optimally excited by the red laser and emits NIR fluorescence maximally at 767 nm.

Bitte beachten Research use only.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: mFluor™ Red 780
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR