EasySep™ Human CD8 Positive Selection Kit II
Lieferant: STEMCELL Technologies
17853RF 17853 100-0699
STMC17853RFEA 1390 EUR
STMC17853RF STMC17853 STMC100-0699
EasySep™ Human CD8 Positive Selection Kit II
Zelltrennmedien
Immunomagnetic positive selection of human CD8+ cells (e.g. CD8+ T cells).
  • Fast and easy-to-use
  • Up to 99% purity
  • No columns required
The EasySep™ Human CD8 Positive Selection Kit II is designed to isolate CD8+ cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic positive selection. Desired cells are targeted with antibody complexes recognising CD8 and magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimise nonspecific binding. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD8 antigen is expressed on cytotoxic T cells and weakly on a subset of NK cells.
This product replaces the EasySep™ Human CD8 Positive Selection Kit (STMC18053) for even faster cell isolations.
Bitte beachten Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).