EasySep™ Mouse CD4+CD62L+ T Cell Isolation Kits
Lieferant: STEMCELL Technologies
18765RF 18765
STMC18765RFEA 2060 EUR
STMC18765RF STMC18765
EasySep™ Mouse CD4+CD62L+ T Cell Isolation Kits
Zelltrennmedien
Immunomagnetic isolation of mouse naïve CD4+ T cells.
  • Fast and easy-to-use
  • Up to 97% purity
  • No columns required
Isolate highly purified naïve CD4+ T cells from single-cell suspensions of splenocytes or other tissues by using a simple, two-step method. First, naive CD4+ T cells are pre-enriched by negative selection using the EasySep™ Mouse Naïve CD4+ T Cell Pre-Enrichment Cocktail containing biotinylated antibodies directed against non-naïve CD4+ T cells (CD8, CD11b, CD11c, CD19, CD24, CD25, CD45R, CD49b, TCRγ/δ, TER119). Then CD4+CD62L+ cells, pre-labelled with CD62L PE, are selected using the EasySep™ PE Selection Cocktail. The desired cells are labelled with antibodies and magnetic particles. The cells are separated without columns using an EasySep™ magnet.
For the isolation of mouse naïve CD4+ T cells in as little as 15 minutes by negative selection, we recommend using the EasySep™ Mouse Naïve CD4+ T Cell Isolation Kit (19765).
Bitte beachten Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').