EasySep™ Human T Cell Isolation Kits
Lieferant: STEMCELL Technologies
17951 17951RF 100-0695
STMC17951EA 1800 EUR
STMC17951 STMC17951RF STMC100-0695
EasySep™ Human T Cell Isolation Kits
Zelltrennmedien
Immunomagnetic isolation of untouched human T cells.
  • Fast, easy-to-use and column-free
  • Up to 98% purity with high recovery
  • Untouched, viable cells
The EasySep™ Human T Cell Isolation Kit is designed to isolate T cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection. The EasySep™ procedure involves labelling unwanted cells with antibody complexes and magnetic particles. The magnetically labelled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
This product can be used in place of the EasySep™ Human T Cell Enrichment Kit for even faster cell isolations.
Bitte beachten Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').