Anti-Histone H3 (di methyl K4) Rabbit Monoclonal Antibody [clone: Y47]

Lieferant: Abcam
AB173324-100 AB173324-1
ABCAAB173324-100EA 1020 EUR
ABCAAB173324-100 ABCAAB173324-1
Anti-Histone H3 (di methyl K4) Rabbit Monoclonal Antibody [clone: Y47]
Antikörper
Rabbit monoclonal [Y47] to Histone H3 (di methyl K4) - BSA and Azide free.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Histone H3
Clonality: Monoclonal
Clone: Y47
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Chicken;Bovine;African green monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR