Anti-Cell adhesion molecule 4 Rabbit Monoclonal Antibody [clone: EP9563]

Lieferant: Abcam
AB248947-100 AB248947-1
ABCAAB248947-100EA 1020 EUR
ABCAAB248947-100 ABCAAB248947-1
Anti-Cell adhesion molecule 4 Rabbit Monoclonal Antibody [clone: EP9563]
Antikörper
Rabbit monoclonal [EP9563] to Cell adhesion molecule 4 - BSA and Azide free.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Cell adhesion molecule 4
Clonality: Monoclonal
Clone: EP9563
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR