Anti-CD13 Rabbit Monoclonal Antibody [clone: EPR20251-123]

Lieferant: Abcam
AB245084-1 AB245084-100
ABCAAB245084-1EA 9320 EUR
ABCAAB245084-1 ABCAAB245084-100
Anti-CD13 Rabbit Monoclonal Antibody [clone: EPR20251-123]
Antikörper
Rabbit monoclonal [EPR20251-123] to CD13 - BSA and Azide free (Detector).

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CD13
Clonality: Monoclonal
Clone: EPR20251-123
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR