Anti-NF-kB p65 Mouse Monoclonal Antibody [clone: E379]

Lieferant: Abcam
AB307841-100 AB307841-1
ABCAAB307841-100EA 1020 EUR
ABCAAB307841-100 ABCAAB307841-1
Anti-NF-kB p65 Mouse Monoclonal Antibody [clone: E379]
Antikörper
Mouse monoclonal [E379] to NF-kB p65 - Chimeric – BSA and Azide free.

Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: NF-kB p65
Clonality: Monoclonal
Clone: E379
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR