Anti-5-methyl cytosine adenosine di-nucleoside (5-mCpA) Mouse Monoclonal Antibody [clone: 2C8H8A6]

Lieferant: Abcam
AB307565-100UG AB307565-20UG AB307565-1MG
ABCAAB307565-100UGEA 694 EUR
ABCAAB307565-100UG ABCAAB307565-20UG ABCAAB307565-1MG
Anti-5-methyl cytosine adenosine di-nucleoside (5-mCpA) Mouse Monoclonal Antibody [clone: 2C8H8A6]
Antikörper
Mouse monoclonal [2C8H8A6] to 5-methyl cytosine adenosine di-nucleoside (5-mCpA).

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: 5-methyl cytosine adenosine di-nucleoside (5-mCpA)
Clonality: Monoclonal
Clone: 2C8H8A6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR