Anti-LMO2 Rabbit Monoclonal Antibody [clone: EP3257]

Lieferant: Abcam
AB247530-100 AB247530-1
ABCAAB247530-100EA 1020 EUR
ABCAAB247530-100 ABCAAB247530-1
Anti-LMO2 Rabbit Monoclonal Antibody [clone: EP3257]
Antikörper
Rabbit monoclonal [EP3257] to LMO2 - BSA and Azide free.

Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: LMO2
Clonality: Monoclonal
Clone: EP3257
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR