Anti-NOR1/TEC Rabbit Monoclonal Antibody [clone: EPR26986-37]

Lieferant: Abcam
AB313781-10UL AB313781-100UL AB313781-20UL AB313781-1ML
ABCAAB313781-10ULEA 0 EUR
ABCAAB313781-10UL ABCAAB313781-100UL ABCAAB313781-20UL ABCAAB313781-1ML
Anti-NOR1/TEC Rabbit Monoclonal Antibody [clone: EPR26986-37]
Antikörper
Rabbit monoclonal [EPR26986-37] to NOR1/TEC.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: NOR1/TEC
Clonality: Monoclonal
Clone: EPR26986-37
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR